3-Pyrrolidinyl-2-phenylacetate hydrochloride

Supplier: Matrix Scientific

049187-500MG 049187-2.5G
101886-652EA 166.15 USD
101886-652 101886-654
3-Pyrrolidinyl-2-phenylacetate hydrochloride
3-Pyrrolidinyl-2-phenylacetate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₂
MW: 241.72 g/mol
MDL Number: MFCD13559738
CAS Number: 1219948-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR