3-Pyrrolidinylmethyl-2-phenylacetate hydrochloride

Supplier: Matrix Scientific

049186-500MG 049186-2.5G
101886-648EA 364.62 USD
101886-648 101886-650
3-Pyrrolidinylmethyl-2-phenylacetate hydrochloride
3-Pyrrolidinylmethyl-2-phenylacetate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₈ClNO₂
MDL Number: MFCD13559737
CAS Number: 1219956-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR