2-(2-Piperidinyl)ethyl-2-phenylacetate hydrochloride

Supplier: Matrix Scientific

049185-500MG 049185-2.5G
101886-644EA 364.62 USD
101886-644 101886-646
2-(2-Piperidinyl)ethyl-2-phenylacetate hydrochloride
2-(2-Piperidinyl)ethyl-2-phenylacetate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MW: 283.8 g/mol
MDL Number: MFCD13559736
CAS Number: 1220020-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR