2-(3-Piperidinyl)ethyl-2-phenylacetate hydrochloride

Supplier: Matrix Scientific

049184-500MG 049184-2.5G
101886-640EA 364.62 USD
101886-640 101886-642
2-(3-Piperidinyl)ethyl-2-phenylacetate hydrochloride
2-(3-Piperidinyl)ethyl-2-phenylacetate hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₂ClNO₂
MDL Number: MFCD13559735
CAS Number: 1219979-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR