3-Azetidinyl-2-phenylacetate

Supplier: Matrix Scientific

049180-500MG 049180-2.5G
101886-624EA 364.62 USD
101886-624 101886-626
3-Azetidinyl-2-phenylacetate
3-Azetidinyl-2-phenylacetate

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD13559731
CAS Number: 1220037-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR