2-[Methyl(2,2,2-trichloroacetyl)amino]acetic acid

Supplier: Matrix Scientific

049176-500MG 049176-2.5G
101886-608EA 364.62 USD
101886-608 101886-610
2-[Methyl(2,2,2-trichloroacetyl)amino]acetic acid
2-[Methyl(2,2,2-trichloroacetyl)amino]acetic acid

Hazard: Irritant

Formula: C₅H₆Cl₃NO₃
MDL Number: MFCD13559727
CAS Number: 1220020-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR