2-Methyl-N-(2,2,2-trichloroacetyl)alanine

Supplier: Matrix Scientific

049175-500MG 049175-2.5G
101886-604EA 364.62 USD
101886-604 101886-606
2-Methyl-N-(2,2,2-trichloroacetyl)alanine
2-Methyl-N-(2,2,2-trichloroacetyl)alanine

Hazard: Irritant

Formula: C₆H₈Cl₃NO₃
MW: 248.49 g/mol
MDL Number: MFCD13559726
CAS Number: 95038-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR