1-(2,2,2-Trichloroacetyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049174-500MG 049174-2.5G
101886-600EA 364.62 USD
101886-600 101886-602
1-(2,2,2-Trichloroacetyl)-4-piperidinecarboxylic acid
1-(2,2,2-Trichloroacetyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₀Cl₃NO₃
MDL Number: MFCD13559725
CAS Number: 1211496-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR