1-(2,2,2-Trichloroacetyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

049173-500MG 049173-2.5G
101886-596EA 364.62 USD
101886-596 101886-598
1-(2,2,2-Trichloroacetyl)-3-piperidinecarboxylic acid
1-(2,2,2-Trichloroacetyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₀Cl₃NO₃
MDL Number: MFCD13559724
CAS Number: 1211491-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR