1-(2,2,2-Trichloroacetyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049172-500MG 049172-2.5G
101886-592EA 364.62 USD
101886-592 101886-594
1-(2,2,2-Trichloroacetyl)-2-piperidinecarboxylic acid
1-(2,2,2-Trichloroacetyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₀Cl₃NO₃
MDL Number: MFCD13559723
CAS Number: 1236256-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR