1-(2,2,2-Trichloroacetyl)proline

Supplier: Matrix Scientific

049171-500MG 049171-2.5G
101886-588EA 364.62 USD
101886-588 101886-590
1-(2,2,2-Trichloroacetyl)proline
1-(2,2,2-Trichloroacetyl)proline

Hazard: Irritant

Formula: C₇H₈Cl₃NO₃
MDL Number: MFCD13559722
CAS Number: 130603-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR