N-(2,2,2-Trichloroacetyl)alanine

Supplier: Matrix Scientific

049170-500MG 049170-2.5G
101886-584EA 364.62 USD
101886-584 101886-586
N-(2,2,2-Trichloroacetyl)alanine
N-(2,2,2-Trichloroacetyl)alanine

Hazard: Irritant

Formula: C₅H₆Cl₃NO₃
MDL Number: MFCD13559721
CAS Number: 5872-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR