2,2,2-Trichloro-N-(4-piperidinyl)acetamide hydrochloride

Supplier: Matrix Scientific

049168-500MG 049168-2.5G
101886-576EA 364.62 USD
101886-576 101886-578
2,2,2-Trichloro-N-(4-piperidinyl)acetamide hydrochloride
2,2,2-Trichloro-N-(4-piperidinyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₂Cl₄N₂O
MDL Number: MFCD13559719
CAS Number: 1219979-92-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR