Methyl (2S,4S)-4-[(2,2,2-trichloroacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049167-500MG 049167-2.5G
101886-572EA 364.62 USD
101886-572 101886-574
Methyl (2S,4S)-4-[(2,2,2-trichloroacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(2,2,2-trichloroacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₈H₁₁Cl₄NO₄
MDL Number: MFCD13559718
CAS Number: 1354485-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR