3-Pyrrolidinyl-2,2,2-trichloroacetate hydrochloride

Supplier: Matrix Scientific

049166-500MG 049166-2.5G
101886-568EA 364.62 USD
101886-568 101886-570
3-Pyrrolidinyl-2,2,2-trichloroacetate hydrochloride
3-Pyrrolidinyl-2,2,2-trichloroacetate hydrochloride

Hazard: Irritant

Formula: C₆H₉Cl₄NO₂
MDL Number: MFCD13559717
CAS Number: 1219948-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR