2-(2-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride

Supplier: Matrix Scientific

049164-500MG 049164-2.5G
101886-560EA 364.62 USD
101886-560 101886-562
2-(2-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride
2-(2-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₅Cl₄NO₂
MDL Number: MFCD13559715
CAS Number: 1220031-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR