2-(3-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride

Supplier: Matrix Scientific

049163-500MG 049163-2.5G
101886-556EA 364.62 USD
101886-556 101886-558
2-(3-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride
2-(3-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₅Cl₄NO₂
MW: 311.04 g/mol
MDL Number: MFCD13559714
CAS Number: 1219979-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR