2-(4-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride
Supplier: Matrix Scientific
049162-500MG
049162-2.5G
101886-552EA
364.62
USD
101886-552
101886-554
2-(4-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride
2-(4-Piperidinyl)ethyl-2,2,2-trichloroacetate hydrochloride
Hazard: Irritant
Formula:
C₉H₁₅Cl₄NO₂ MW: 311.04 g/mol |
MDL Number:
MFCD13559713 CAS Number: 1219979-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...