4-Piperidinylmethyl-2,2,2-trichloroacetate hydrochloride

Supplier: Matrix Scientific

049161-500MG 049161-2.5G
101886-548EA 364.62 USD
101886-548 101886-550
4-Piperidinylmethyl-2,2,2-trichloroacetate hydrochloride
4-Piperidinylmethyl-2,2,2-trichloroacetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₃Cl₄NO₂
MW: 297.01 g/mol
MDL Number: MFCD13559712
CAS Number: 1220020-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR