3-Azetidinyl-2,2,2-trichloroacetate

Supplier: Matrix Scientific

049159-500MG 049159-2.5G
101886-540EA 364.62 USD
101886-540 101886-542
3-Azetidinyl-2,2,2-trichloroacetate
3-Azetidinyl-2,2,2-trichloroacetate

Hazard: Irritant

Formula: C₅H₆Cl₃NO₂
MDL Number: MFCD13559710
CAS Number: 1219956-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR