4-Piperidinyl-2,2,2-trichloroacetate hydrochloride
Supplier: Matrix Scientific
049158-500MG
049158-2.5G
101886-536EA
364.62
USD
101886-536
101886-538
4-Piperidinyl-2,2,2-trichloroacetate hydrochloride
4-Piperidinyl-2,2,2-trichloroacetate hydrochloride
Hazard: Irritant
Formula:
C₇H₁₁Cl₄NO₂ |
MDL Number:
MFCD13559709 CAS Number: 1219948-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...