3-Piperidinyl-2,2,2-trichloroacetate hydrochloride
Supplier: Matrix Scientific
049157-500MG
049157-2.5G
101886-532EA
364.62
USD
101886-532
101886-534
3-Piperidinyl-2,2,2-trichloroacetate hydrochloride
3-Piperidinyl-2,2,2-trichloroacetate hydrochloride
Hazard: Irritant
Formula:
C₇H₁₁Cl₄NO₂ |
MDL Number:
MFCD13559708 CAS Number: 1219979-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...