2-[(2-Chloroacetyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

049155-500MG 049155-2.5G
101886-524EA 186.15 USD
101886-524 101886-526
2-[(2-Chloroacetyl)(methyl)amino]acetic acid
2-[(2-Chloroacetyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₅H₈ClNO₃
MDL Number: MFCD13559707
CAS Number: 65332-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR