N-(2-Chloroacetyl)-2-methylalanine

Supplier: Matrix Scientific

049154-500MG 049154-2.5G
101886-520EA 253.85 USD
101886-520 101886-522
N-(2-Chloroacetyl)-2-methylalanine
N-(2-Chloroacetyl)-2-methylalanine

Hazard: Irritant

Formula: C₆H₁₀ClNO₃
MDL Number: MFCD13559706
CAS Number: 95038-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR