1-(2-Chloroacetyl)-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

049153-500MG 049153-2.5G
101886-516EA 364.62 USD
101886-516 101886-518
1-(2-Chloroacetyl)-4-piperidinecarboxylic acid
1-(2-Chloroacetyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₂ClNO₃
MDL Number: MFCD02093986
CAS Number: 318280-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR