1-(2-Chloroacetyl)-3-piperidinecarboxylic acid
Supplier: Matrix Scientific
049152-500MG
049152-2.5G
101886-512EA
364.62
USD
101886-512
101886-514
1-(2-Chloroacetyl)-3-piperidinecarboxylic acid
1-(2-Chloroacetyl)-3-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₈H₁₂ClNO₃ |
MDL Number:
MFCD13559705 CAS Number: 1219948-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...