1-(2-Chloroacetyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049151-500MG 049151-2.5G
101886-508EA 364.62 USD
101886-508 101886-510
1-(2-Chloroacetyl)-2-piperidinecarboxylic acid
1-(2-Chloroacetyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₂ClNO₃
MDL Number: MFCD13559704
CAS Number: 1236257-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR