N-Chloroacetyl-DL-alanine

Supplier: Matrix Scientific

049149-500MG 049149-2.5G
101886-500EA 364.62 USD
101886-500 101886-502
N-Chloroacetyl-DL-alanine
N-Chloroacetyl-DL-alanine

Hazard: Irritant

Formula: C₅H₈ClNO₃
MDL Number: MFCD00020401
CAS Number: 1190-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR