N-Chloroacetylglycine

Supplier: Matrix Scientific

Synonyms: Chloroac-Gly-OH, N-Chloroacetylglycine, 2-(2-Chloroacetamido)acetic acid

049148-500MG 049148-2.5G
101886-496EA 364.62 USD
101886-496 101886-498
N-Chloroacetylglycine
N-Chloroacetylglycine

Hazard: Irritant

Formula: C₄H₆ClNO₃
MW: 151.55 g/mol
MDL Number: MFCD00021733
CAS Number: 6319-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR