2-Chloro-N-(4-piperidinyl)acetamide hydrochloride

Supplier: Matrix Scientific

049147-500MG 049147-2.5G
101886-492EA 364.62 USD
101886-492 101886-494
2-Chloro-N-(4-piperidinyl)acetamide hydrochloride
2-Chloro-N-(4-piperidinyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₇H₁₄Cl₂N₂O
MDL Number: MFCD13559703
CAS Number: 1220020-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR