3-Pyrrolidinylmethyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049144-500MG 049144-2.5G
101886-480EA 364.62 USD
101886-480 101886-482
3-Pyrrolidinylmethyl-2-chloroacetate hydrochloride
3-Pyrrolidinylmethyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₇H₁₃Cl₂NO₂
MDL Number: MFCD13559700
CAS Number: 1219979-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR