2-(2-Piperidinyl)ethyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049143-500MG 049143-2.5G
101886-476EA 364.62 USD
101886-476 101886-478
2-(2-Piperidinyl)ethyl-2-chloroacetate hydrochloride
2-(2-Piperidinyl)ethyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₇Cl₂NO₂
MDL Number: MFCD13559699
CAS Number: 1219956-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR