4-Piperidinylmethyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049140-500MG 049140-2.5G
101886-464EA 364.62 USD
101886-464 101886-466
4-Piperidinylmethyl-2-chloroacetate hydrochloride
4-Piperidinylmethyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₈H₁₅Cl₂NO₂
MDL Number: MFCD13559696
CAS Number: 1219949-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR