3-Azetidinyl-2-chloroacetate

Supplier: Matrix Scientific

049138-500MG 049138-2.5G
101886-456EA 364.62 USD
101886-456 101886-458
3-Azetidinyl-2-chloroacetate
3-Azetidinyl-2-chloroacetate

Hazard: Irritant

Formula: C₅H₈ClNO₂
MDL Number: MFCD13559694
CAS Number: 1220020-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR