4-Piperidinyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049137-500MG 049137-2.5G
101886-452EA 364.62 USD
101886-452 101886-454
4-Piperidinyl-2-chloroacetate hydrochloride
4-Piperidinyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₇H₁₃Cl₂NO₂
MW: 214.09 g/mol
MDL Number: MFCD13559693
CAS Number: 1220020-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR