3-Piperidinyl-2-chloroacetate hydrochloride

Supplier: Matrix Scientific

049136-500MG 049136-2.5G
101886-448EA 364.62 USD
101886-448 101886-450
3-Piperidinyl-2-chloroacetate hydrochloride
3-Piperidinyl-2-chloroacetate hydrochloride

Hazard: Irritant

Formula: C₇H₁₃Cl₂NO₂
MDL Number: MFCD13559692
CAS Number: 1220037-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR