N-(2-Methoxyacetyl)-β-alanine

Supplier: Matrix Scientific

049135-500MG 049135-2.5G
101886-444EA 364.62 USD
101886-444 101886-446
N-(2-Methoxyacetyl)-β-alanine
N-(2-Methoxyacetyl)-beta-alanine

Hazard: Irritant

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
MDL Number: MFCD09050610
CAS Number: 36855-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR