2-[(2-Methoxyacetyl)(methyl)amino]acetic acid

Supplier: Matrix Scientific

049134-500MG 049134-2.5G
101886-440EA 384.62 USD
101886-440 101886-442
2-[(2-Methoxyacetyl)(methyl)amino]acetic acid
2-[(2-Methoxyacetyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₆H₁₁NO₄
MDL Number: MFCD09049704
CAS Number: 926228-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR