1-(2-Methoxyacetyl)-2-piperidinecarboxylic acid

Supplier: Matrix Scientific

049132-500MG 049132-2.5G
101886-432EA 364.62 USD
101886-432 101886-434
1-(2-Methoxyacetyl)-2-piperidinecarboxylic acid
1-(2-Methoxyacetyl)-2-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₅NO₄
MDL Number: MFCD09048303
CAS Number: 1101744-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR