1-(2-Methoxyacetyl)proline

Supplier: Matrix Scientific

049131-500MG 049131-2.5G
101886-428EA 364.62 USD
101886-428 101886-430
1-(2-Methoxyacetyl)proline
1-(2-Methoxyacetyl)proline

Hazard: Irritant

Formula: C₈H₁₃NO₄
MDL Number: MFCD09046233
CAS Number: 130603-11-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR