N-(2-Methoxyacetyl)alanine

Supplier: Matrix Scientific

049130-500MG 049130-2.5G
101886-424EA 384.62 USD
101886-424 101886-426
N-(2-Methoxyacetyl)alanine
N-(2-Methoxyacetyl)alanine

Hazard: Irritant

Formula: C₆H₁₁NO₄
MDL Number: MFCD09046076
CAS Number: 257638-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR