2-[(2-Methoxyacetyl)amino]acetic acid

Supplier: Matrix Scientific

049129-500MG 049129-2.5G
101886-420EA 186.15 USD
101886-420 101886-422
2-[(2-Methoxyacetyl)amino]acetic acid
2-[(2-Methoxyacetyl)amino]acetic acid

Hazard: Irritant

Formula: C₅H₉NO₄
MDL Number: MFCD09048365
CAS Number: 97871-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR