2-Methoxy-N-(4-piperidinyl)acetamide hydrochloride
Supplier: Matrix Scientific
049128-500MG
049128-2.5G
101886-416EA
364.62
USD
101886-416
101886-418
2-Methoxy-N-(4-piperidinyl)acetamide hydrochloride
2-Methoxy-N-(4-piperidinyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₇ClN₂O₂ |
MDL Number:
MFCD11809617 CAS Number: 1170286-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...