Methyl (2S,4S)-4-[(2-methoxyacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Supplier: Matrix Scientific

049127-500MG 049127-2.5G
101886-412EA 364.62 USD
101886-412 101886-414
Methyl (2S,4S)-4-[(2-methoxyacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride
Methyl (2S,4S)-4-[(2-methoxyacetyl)oxy]-2-pyrrolidinecarboxylate hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₅
MDL Number: MFCD13559690
CAS Number: 1354490-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR