3-Pyrrolidinyl-2-methoxyacetate hydrochloride

Supplier: Matrix Scientific

049126-500MG 049126-2.5G
101886-408EA 364.62 USD
101886-408 101886-410
3-Pyrrolidinyl-2-methoxyacetate hydrochloride
3-Pyrrolidinyl-2-methoxyacetate hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₃
MDL Number: MFCD13559689
CAS Number: 1220037-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR