2-(2-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
Supplier: Matrix Scientific
049124-500MG
049124-2.5G
101886-400EA
364.62
USD
101886-400
101886-402
2-(2-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
2-(2-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀ClNO₃ |
MDL Number:
MFCD13559687 CAS Number: 1220031-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...