2-(3-Piperidinyl)ethyl-2-methoxyacetate hydrochloride

Supplier: Matrix Scientific

049123-500MG 049123-2.5G
101886-396EA 364.62 USD
101886-396 101886-398
2-(3-Piperidinyl)ethyl-2-methoxyacetate hydrochloride
2-(3-Piperidinyl)ethyl-2-methoxyacetate hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₀ClNO₃
MW: 237.73 g/mol
MDL Number: MFCD13559686
CAS Number: 1220020-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR