3-Piperidinylmethyl-2-methoxyacetate hydrochloride

Supplier: Matrix Scientific

049120-500MG 049120-2.5G
101886-384EA 364.62 USD
101886-384 101886-386
3-Piperidinylmethyl-2-methoxyacetate hydrochloride
3-Piperidinylmethyl-2-methoxyacetate hydrochloride

Hazard: Irritant

Formula: C₉H₁₈ClNO₃
MW: 223.7 g/mol
MDL Number: MFCD13559683
CAS Number: 1219979-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR