3-Azetidinyl-2-methoxyacetate

Supplier: Matrix Scientific

049119-500MG 049119-2.5G
101886-380EA 364.62 USD
101886-380 101886-382
3-Azetidinyl-2-methoxyacetate
3-Azetidinyl-2-methoxyacetate

Hazard: Irritant

Formula: C₆H₁₁NO₃
MDL Number: MFCD13559682
CAS Number: 1219949-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR