4-Piperidinyl-2-methoxyacetate hydrochloride
Supplier: Matrix Scientific
049118-500MG
049118-2.5G
101886-376EA
364.62
USD
101886-376
101886-378
4-Piperidinyl-2-methoxyacetate hydrochloride
4-Piperidinyl-2-methoxyacetate hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClNO₃ MW: 209.67 g/mol |
MDL Number:
MFCD13559681 CAS Number: 1220031-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...